CHIS5-05229

5-05230 5-05229
76228-328EA 278.41 USD
76228-328 76228-330
CHIS5-05229
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR