CHIS5-05221

5-05222 5-05221
76228-314EA 524.06 USD
76228-314 76228-312
CHIS5-05221
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR