CHIS5-05205

5-05206 5-05205
76229-712EA 22.93 USD
76229-712 76229-714
CHIS5-05205
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR