CHIS5-05187

5-05188 5-05187
76228-278EA 979.34 USD
76228-278 76228-276
CHIS5-05187
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR