CHIS5-05183

5-05183 5-05184
76228-270EA 1555.82 USD
76228-270 76228-268
CHIS5-05183
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR