CHIS5-05177

5-05178 5-05177
76228-258EA 655.08 USD
76228-258 76228-256
CHIS5-05177
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR