CHIS5-05175

5-05175 5-05176
76228-254EA 517.51 USD
76228-254 76228-252
CHIS5-05175
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR