CHIS5-05151

5-05152 5-05151
76228-206EA 1203.71 USD
76228-206 76228-204
CHIS5-05151
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR