CHIS5-05147

5-05147 5-05148
76228-198EA 818.85 USD
76228-198 76228-196
CHIS5-05147
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR