CHIS5-05145

5-05146 5-05145
76228-192EA 70.43 USD
76228-192 76228-194
CHIS5-05145
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR