CHIS5-05095

5-05096 5-05095
76228-102EA 414.33 USD
76228-102 76228-100
CHIS5-05095
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR