CHIS5-05065

5-05065 5-05066
76228-042EA 171.95 USD
76228-042 76228-040
CHIS5-05065
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR