CHIS5-05049

5-05049 5-05050
76228-008EA 131.02 USD
76228-008 76228-010
CHIS5-05049
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR