CHIS5-05017

5-05018 5-05017
76227-960EA 52.41 USD
76227-960 76227-962
CHIS5-05017
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR