CHIS5-04991

5-04991 5-04992
76227-908EA 139.21 USD
76227-908 76227-910
CHIS5-04991
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR