CHIS5-04987

5-04987 5-04988
76227-900EA 180.15 USD
76227-900 76227-902
CHIS5-04987
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR