CHIS5-04977

5-04977 5-04978
76227-880EA 131.02 USD
76227-880 76227-882
CHIS5-04977
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR