CHIS5-04897

5-04897 5-04898
76229-682EA 786.1 USD
76229-682 76229-680
CHIS5-04897
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR