CHIS5-04893

5-04893 5-04894
76229-674EA 196.52 USD
76229-674 76229-672
CHIS5-04893
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR