CHIS5-04887

5-04888 5-04887
76227-740EA 189.97 USD
76227-740 76227-742
CHIS5-04887
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR