CHIS5-04815

5-04815 5-04816
76227-604EA 196.52 USD
76227-604 76227-606
CHIS5-04815
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR