CHIS5-04779

5-04779 5-04780
76227-532EA 131.02 USD
76227-532 76227-534
CHIS5-04779
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR