CHIS5-04717

5-04718 5-04717
76227-416EA 122.83 USD
76227-416 76227-418
CHIS5-04717
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR