CHIS5-04715

5-04716 5-04715
76227-412EA 196.52 USD
76227-412 76227-414
CHIS5-04715
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR