CHIS5-04713

5-04713 5-04714
76227-410EA 394.69 USD
76227-410 76227-408
CHIS5-04713
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR