CHIS5-04697

5-04697 5-04698
76229-624EA 393.05 USD
76229-624 76229-626
CHIS5-04697
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR