CHIS5-04673

5-04674 5-04673
76227-360EA 81.89 USD
76227-360 76227-362
CHIS5-04673
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR