CHIS5-04667

5-04668 5-04667
76227-350EA 999 USD
76227-350 76227-348
CHIS5-04667
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR