CHIS5-04655

5-04656 5-04655
76227-324EA 294.79 USD
76227-324 76227-326
CHIS5-04655
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR