CHIS5-04623

5-04623 5-04624
76227-284EA 163.77 USD
76227-284 76227-286
CHIS5-04623
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR