CHIS5-04617

5-04617 5-04618
76227-272EA 589.57 USD
76227-272 76227-274
CHIS5-04617
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR