CHIS5-04569

5-04569 5-04570
76227-184EA 57.32 USD
76227-184 76227-186
CHIS5-04569
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR