CHIS5-04533

5-04533 5-04534
76227-130EA 704.21 USD
76227-130 76227-128
CHIS5-04533
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR