CHIS5-04497

5-04498 5-04497
76229-568EA 237.47 USD
76229-568 76229-570
CHIS5-04497
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR