CHIS5-04491

5-04491 5-04492
76229-558EA 294.79 USD
76229-558 76229-556
CHIS5-04491
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR