CHIS5-04489

5-04490 5-04489
76227-056EA 278.41 USD
76227-056 76227-058
CHIS5-04489
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR