CHIS5-04479

5-04479 5-04480
76227-036EA 327.54 USD
76227-036 76227-038
CHIS5-04479
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR