CHIS5-04475

5-04475 5-04476
76226-356EA 343.92 USD
76226-356 76229-550
CHIS5-04475
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR