CHIS5-04469

5-04469 5-04470
76227-032EA 262.03 USD
76227-032 76227-034
CHIS5-04469
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR