CHIS5-04457

5-04458 5-04457
76227-016EA 311.16 USD
76227-016 76227-018
CHIS5-04457
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR