CHIS5-04455

5-04456 5-04455
76227-012EA 294.79 USD
76227-012 76227-014
CHIS5-04455
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR