CHIS5-04453

5-04453 5-04454
76227-010EA 1064.51 USD
76227-010 76227-008
CHIS5-04453
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR