CHIS5-04437

5-04437 5-04438
76226-320EA 327.54 USD
76226-320 76226-322
CHIS5-04437
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR