CHIS5-04419

5-04420 5-04419
76226-980EA 327.54 USD
76226-980 76226-982
CHIS5-04419
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR