CHIS5-04381

5-04381 5-04382
76229-504EA 212.9 USD
76229-504 76229-506
CHIS5-04381
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR