CHIS5-04379

5-04379 5-04380
76229-502EA 818.85 USD
76229-502 76229-500
CHIS5-04379
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR