CHIS5-04371

5-04371 5-04372
76229-484EA 253.84 USD
76229-484 76229-486
CHIS5-04371
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR