CHIS5-04357

5-04358 5-04357
76229-458EA 818.85 USD
76229-458 76229-456
CHIS5-04357
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR