CHIS5-04309

5-04310 5-04309
76226-896EA 491.31 USD
76226-896 76226-898
CHIS5-04309
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR