CHIS5-04293

5-04293 5-04294
76226-874EA 1392.05 USD
76226-874 76226-872
CHIS5-04293
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR